siRNA Details
Gene Symbol: CARD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052889.2 | NP_443121.1 | 2, 3 | 309 | ||
GenBank mRNA | AF367017.1 | - | 1, 2 | 282 | ||
AK311902.1 | - | 2, 3 | 318 | |||
BC070196.1 | - | 2, 3 | 309 | |||
BC104635.1 | - | 2, 3 | 313 | |||
BC117478.1 | - | 2, 3 | 361 | |||
BC117480.1 | - | 2, 3 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
