siRNA Details
Gene Symbol: Usp17ld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001559.2 | NP_001001559.2 | 2 | 749 | ||
GenBank mRNA | AF393638.1 | - | 2 | 749 |
Gene Symbol: Usp17lc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010089.3 | NP_034219.3 | 771 | |||
GenBank mRNA | U70369.1 | - | 771 |
Gene Symbol: Usp17le | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256973.1 | NP_001243902.1 | 3 | 851 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
