siRNA Details
Gene Symbol: Lefty1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010094.3 | NP_034224.1 | 3, 4 | 814 | ||
GenBank mRNA | AJ000082.1 | - | 3, 4 | 794 | ||
AK131943.1 | - | 3, 4 | 816 | |||
BC050221.1 | - | 3, 4 | 777 | 30005856 | ||
D83921.1 | - | 3, 4 | 831 | |||
Z73151.1 | - | 3, 4 | 794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
