siRNA Details
Gene Symbol: Ebf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113414.1 | NP_001106885.1 | 7, 8 | 1169 | ||
NM_001113415.1 | NP_001106886.1 | 7, 8 | 1169 | |||
NM_010096.3 | NP_034226.1 | 7, 8 | 1169 | |||
GenBank mRNA | AK138602.1 | - | 2, 3 | 358 | ||
AK220542.1 | - | 7, 8 | 1169 | |||
BC067018.1 | - | 8, 9 | 924 | 6833120 | ||
CF727275.1 | - | 1, 2 | 34 | |||
U82442.1 | - | 6, 7 | 466 | |||
U92702.1 | - | 7, 8 | 710 | |||
U92704.1 | - | 7, 8 | 710 |

More Information
Other Species Targeted

Gene Ontology Categories:
