siRNA Details
Gene Symbol: GATA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080473.4 | NP_536721.1 | 5 | 922 | ||
XM_006723699.3 | XP_006723762.1 | 5 | 942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
