siRNA Details
Gene Symbol: Fem1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010192.4 | NP_034322.3 | 1 | 1809 | ||
GenBank mRNA | AK005041.1 | - | 1 | 1768 | ||
AK009354.1 | - | 1 | 102 | |||
AK034412.1 | - | 1 | 1784 | |||
AK080978.1 | - | 1 | 1781 | |||
AK129000.1 | - | 1 | 1792 | |||
AK147879.1 | - | 1 | 1794 | |||
AK155363.1 | - | 1 | 1793 | |||
BC009161.1 | - | 1 | 1776 | 2811079 | ||
BC054382.1 | - | 1 | 1780 | 6469158 | ||
BC110669.1 | - | 1 | 1763 | 5042239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
