siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079908.2 | NP_001073377.1 | 4 | 1178 | ||
NM_001079909.2 | NP_001073378.1 | 3 | 911 | |||
NM_010206.3 | NP_034336.2 | 4 | 1178 | |||
GenBank mRNA | AF176552.1 | - | 2 | 209 | ||
AK028354.1 | - | 3 | 351 | |||
AK049704.1 | - | 4 | 1146 | |||
BC010200.1 | - | 4 | 1077 | 3967874 | ||
BC033447.1 | - | 2 | 180 | 4037899 | ||
M28998.1 | - | 3 | 476 | |||
M33760.1 | - | 2 | 195 | |||
M65053.1 | - | 3 | 455 | |||
U22324.1 | - | 3 | 476 | |||
U23445.1 | - | 2 | 209 | |||
X51893.1 | - | 4 | 539 | |||
X57790.1 | - | 3 | 419 |
