siRNA Details
Gene Symbol: Eya3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010166.3 | NP_034296.2 | 7 | 550 | ||
NM_210071.2 | NP_997592.1 | 6 | 511 | |||
GenBank mRNA | AK144653.1 | - | 6 | 502 | ||
AK164591.1 | - | 6 | 511 | |||
BC063259.1 | - | 6 | 439 | 6406090 | ||
BY730487.1 | - | 6 | 511 | |||
U81604.1 | - | 7 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
