siRNA Details
Gene Symbol: Fgfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010207.2 | NP_034337.2 | 3 | 1518 | ||
GenBank mRNA | AK038588.1 | - | 3 | 935 | ||
BC072640.1 | - | 3 | 955 | |||
EF143323.1 | - | 2 | 294 | |||
EF143328.1 | - | 2 | 294 | |||
EF143329.1 | - | 2 | 294 | |||
EF143335.1 | - | 2 | 294 | |||
EF143338.1 | - | 2 | 294 | |||
M86441.1 | - | 3 | 900 | |||
X55441.1 | - | 3 | 951 |

More Information
Other Species Targeted

Gene Ontology Categories:
