siRNA Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017861.1 | - | 3 | 358 | ||
AK146459.1 | - | 4 | 474 | |||
AK158704.1 | - | 4 | 476 | |||
BC008130.1 | - | 5 | 479 | 3498984 | ||
BC034861.1 | - | 4 | 483 | |||
X71978.1 | - | 4 | 498 | |||
Z67963.1 | - | 4 | 498 |

More Information
Other Species Targeted

