siRNA Details
Gene Symbol: Fcgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010188.5 | NP_034318.2 | 5 | 1210 | ||
GenBank mRNA | AK077227.1 | - | 5 | 1170 | ||
AK151557.1 | - | 5 | 1170 | |||
AK151795.1 | - | 5 | 1169 | |||
AK152875.1 | - | 5 | 1170 | |||
AK153384.1 | - | 5 | 1169 | |||
AK153449.1 | - | 5 | 1169 | |||
AK154892.1 | - | 5 | 1171 | |||
AK159919.1 | - | 5 | 1156 | |||
AK163401.1 | - | 3 | 1160 | |||
AK169887.1 | - | 5 | 1160 | |||
AK171289.1 | - | 5 | 1164 | |||
BC052819.1 | - | 5 | 1164 | 30044058 | ||
M14215.1 | - | 5 | 1197 |
