siRNA Details
Gene Symbol: Fem1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010192.4 | NP_034322.3 | 1 | 2436 | ||
GenBank mRNA | AK009354.1 | - | 1 | 729 | ||
AK034412.1 | - | 1 | 2411 | |||
AK075860.1 | - | 1 | 560 | |||
AK080978.1 | - | 1 | 2408 | |||
AK129000.1 | - | 1 | 2419 | |||
AK147879.1 | - | 1 | 2421 | |||
AK155363.1 | - | 1 | 2420 | |||
AK205548.1 | - | 1 | 191 | |||
BC009161.1 | - | 1 | 2404 | 2811079 | ||
BC054382.1 | - | 1 | 2407 | 6469158 | ||
BC110669.1 | - | 1 | 2390 | 5042239 |
Gene Symbol: 4931440F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176829.2 | NP_789799.1 | 1 | 2532 | ||
GenBank mRNA | AK029918.1 | - | 1 | 2532 | ||
BC139107.1 | - | 1 | 2389 | 8862129 | ||
BC139123.1 | - | 1 | 2389 | 8862145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
