siRNA Details
Gene Symbol: Fgfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010207.2 | NP_034337.2 | 8 | 2207 | ||
GenBank mRNA | BC065069.1 | - | 6 | 1293 | ||
BC151201.1 | - | 6 | 1067 | 9088089 | ||
EF143332.1 | - | 7 | 864 | |||
EF143333.1 | - | 3 | 344 | |||
EF143334.1 | - | 5 | 638 | |||
EF143335.1 | - | 7 | 983 | |||
EF143338.1 | - | 7 | 983 | |||
EF143339.1 | - | 5 | 638 | |||
EF143340.1 | - | 5 | 638 | |||
M86441.1 | - | 8 | 1589 | |||
X55441.1 | - | 8 | 1637 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109892, NM_001109894, NM_001109896, XM_006230390, XM_006230393, and others.
Gene Ontology Categories:
