siRNA Details
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145820.1 | NP_001139292.1 | 12, 13 | 1935 | ||
NM_010274.3 | NP_034404.3 | 12, 13 | 1810 | |||
GenBank mRNA | AK034353.1 | - | 12, 13 | 1719 | ||
AK144716.1 | - | 12, 13 | 1936 | |||
AK151189.1 | - | 12, 13 | 1723 | |||
AK167152.1 | - | 12, 13 | 1748 | |||
D50430.1 | - | 12, 13 | 1712 | |||
U60987.1 | - | 12, 13 | 1746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
