siRNA Details
Gene Symbol: OR7E91P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002185.3 | 3 | 915 | |||
GenBank mRNA | BC014374.1 | - | 3 | 918 | ||
BC030991.1 | - | 3 | 954 |
Gene Symbol: OR7E125P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF359416.1 | - | LIT2085 PJCG6 pseudogene |
1 | 337 |
Gene Symbol: OR7E154P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122996.1 | - | 1 | 4260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
