siRNA Details
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010387.3 | NP_034517.2 | 2 | 449 | ||
GenBank mRNA | AK032745.1 | - | 2 | 450 | ||
AK169969.1 | - | 2 | 430 | |||
AK170866.1 | - | 2 | 432 | |||
BC002237.1 | - | 411 | 3491702 | |||
BC003718.1 | - | 2 | 389 | 3486844 | ||
KJ650225.1 | - | 2 | 241 | |||
U20309.1 | - | 1 | 187 | |||
U20310.1 | - | 1 | 187 | |||
U20312.1 | - | 1 | 187 | |||
U35329.1 | - | 241 | ||||
U35330.1 | - | 241 | ||||
U35331.1 | - | 241 | ||||
U35332.1 | - | 241 | ||||
U35333.1 | - | 241 | ||||
X62743.1 | - | 349 | ||||
X90964.1 | - | 241 | ||||
X90965.1 | - | 241 | ||||
X90966.1 | - | 241 | ||||
X90967.1 | - | 241 | ||||
X90968.1 | - | 241 | ||||
X90969.1 | - | 241 | ||||
X90970.1 | - | 241 | ||||
X90971.1 | - | 241 | ||||
X90972.1 | - | 241 | ||||
X90973.1 | - | 241 | ||||
X90974.1 | - | 241 | ||||
X90975.1 | - | 241 |
