siRNA Details
Gene Symbol: H2-DMb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010388.4 | NP_034518.1 | 2 | 257 | ||
GenBank mRNA | AK154923.1 | - | 2 | 361 | ||
BC052864.1 | - | 3 | 512 | |||
KJ650224.1 | - | 2 | 257 | |||
U35334.1 | - | 2 | 257 | |||
U35335.1 | - | 2 | 257 | |||
X90802.1 | - | 2 | 257 | |||
X90804.1 | - | 2 | 257 |
