siRNA Details
Gene Symbol: H2-Q7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198560.1 | NP_001185489.1 | 4 | 718 | ||
NM_010394.4 | NP_034524.3 | 4 | 718 | |||
GenBank mRNA | BC012038.1 | - | 1 | 121 | ||
BC090638.1 | - | 3 | 2748 | |||
X05389.1 | - | 652 |
Gene Symbol: H2-Q6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207648.1 | NP_997531.1 | 4 | 652 |
Gene Symbol: H2-Q8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023124.5 | NP_075613.3 | 4 | 680 |
Gene Symbol: H2-Q9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201460.1 | NP_001188389.1 | 4 | 706 | ||
GenBank mRNA | AY989883.1 | - | 4 | 657 | ||
BC120554.1 | - | 4 | 718 | 40129477 | ||
CN717653.1 | - | 2 | 277 |
Gene Symbol: LOC68395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025170.1 | - | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
