siRNA Details
Gene Symbol: Hdac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077696.1 | NP_001071164.1 | 5, 6 | 862 | ||
NM_001284248.1 | NP_001271177.1 | 7, 8 | 1374 | |||
NM_001284249.1 | NP_001271178.1 | 6, 7 | 971 | |||
NM_001284250.1 | NP_001271179.1 | 5, 6 | 859 | |||
NM_010412.3 | NP_034542.3 | 5, 6 | 859 | |||
GenBank mRNA | AF006602.2 | - | 4, 5 | 575 | ||
AF207748.1 | - | 4, 5 | 494 | |||
AK139202.1 | - | 6, 7 | 944 | |||
AK145972.1 | - | 5, 6 | 825 | |||
AK146483.1 | - | 5, 6 | 798 | |||
AK155335.1 | - | 5, 6 | 833 | |||
AK156008.1 | - | 3 | 1017 | |||
AK157159.1 | - | 3 | 1017 | |||
BC049926.1 | - | 5, 6 | 795 | |||
BC058554.1 | - | 7, 8 | 1326 | |||
BC060609.1 | - | 4, 5 | 653 | 5720438 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053450.
Gene Ontology Categories:
