siRNA Details
Gene Symbol: PTPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253829.1 | NP_001240758.1 | 4, 5 | 719 | ||
NM_001253830.1 | NP_001240759.1 | 5, 6 | 780 | |||
NM_152422.4 | NP_689635.3 | 4, 5 | 713 | |||
NM_177995.2 | NP_818931.1 | 5, 6 | 795 | |||
XM_005251711.4 | XP_005251768.1 | 5, 6 | 2737 | |||
XM_011518240.2 | XP_011516542.1 | 5, 6 | 545 | |||
XM_011518241.2 | XP_011516543.1 | 5, 6 | 499 | |||
XM_017014280.1 | XP_016869769.1 | 6, 7 | 604 | |||
XM_017014281.1 | XP_016869770.1 | 5, 6 | 523 | |||
GenBank mRNA | AK124912.1 | - | 4, 5 | 599 | ||
AK289692.1 | - | 4, 5 | 670 | |||
AK310403.1 | - | 4, 5 | 677 | |||
AY171233.1 | - | 5, 6 | 800 | |||
BC067120.1 | - | 5, 6 | 780 | 5539182 | ||
DA312531.1 | - | 3, 4 | 295 |
