siRNA Details
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169969.1 | - | 5, 6 | 972 | ||
AK170866.1 | - | 5, 6 | 974 | |||
BC002237.1 | - | 953 | 3491702 | |||
X62743.1 | - | 891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
