siRNA Details
Gene Symbol: OR7E5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027688.2 | 4 | 713 | |||
GenBank mRNA | AK055955.1 | - | 4 | 713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
