siRNA Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111274.1 | NP_001104744.1 | 4, 5 | 663 | ||
NM_010512.4 | NP_034642.2 | 4, 5 | 745 | |||
GenBank mRNA | AY878193.1 | - | 4, 5 | 543 | ||
BC012409.1 | - | 3, 4 | 400 | 4194295 | ||
CT010364.1 | - | 3, 4 | 373 | |||
X04482.1 | - | 4, 5 | 445 |

More Information
Other Species Targeted

Gene Ontology Categories:
