siRNA Details
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111320.1 | NP_001104790.1 | 3 | 385 | ||
NM_010497.3 | NP_034627.3 | 3 | 363 | |||
GenBank mRNA | AF020039.1 | - | 2 | 213 | ||
AK087063.1 | - | 3 | 265 | |||
AK149019.1 | - | 3 | 294 | |||
AK151212.1 | - | 3 | 288 | |||
AK159173.1 | - | 3 | 294 | |||
AK160896.1 | - | 3 | 295 | |||
AK167158.1 | - | 3 | 335 | |||
AK167583.1 | - | 3 | 299 | |||
AK168832.1 | - | 3 | 337 | |||
CF950262.1 | - | 3 | 324 | |||
DQ403075.1 | - | 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
