siRNA Details
Gene Symbol: Hira | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010435.2 | NP_034565.2 | 14, 15 | 2346 | ||
GenBank mRNA | BC060181.1 | - | 14, 15 | 2030 | ||
X75295.1 | - | 15, 16 | 1861 | |||
X92590.1 | - | 14, 15 | 1833 | |||
X99712.1 | - | 14, 15 | 1673 |

More Information
Other Species Targeted

Gene Ontology Categories:
