siRNA Details
Gene Symbol: H2afx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010436.2 | NP_034566.1 | 1 | 1343 | ||
GenBank mRNA | AK088040.1 | - | 1 | 1365 | ||
AK217455.1 | - | 1 | 118 | |||
BC005468.1 | - | 1 | 1337 | 3490058 | ||
BC010336.1 | - | 1 | 1343 | 3156946 | ||
X58069.1 | - | 1 | 1345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
