siRNA Details
Gene Symbol: Hmox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136066.2 | NP_001129538.1 | 6 | 1186 | ||
NM_010443.2 | NP_034573.2 | 6 | 1202 | |||
GenBank mRNA | AF029874.1 | - | 6 | 1199 | ||
AF054669.1 | - | 6 | 1203 | |||
AF054670.1 | - | 6 | 1137 | |||
AK032175.1 | - | 6 | 1144 | |||
AK134536.1 | - | 6 | 1161 | |||
AK152936.1 | - | 6 | 1124 | |||
BC002011.1 | - | 6 | 1158 | 3484259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
