siRNA Details
Gene Symbol: Hoxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010451.2 | NP_034581.1 | 2 | 925 | ||
GenBank mRNA | AB221621.1 | - | 2 | 742 | ||
AK134501.1 | - | 2 | 574 | |||
AK157713.1 | - | 2 | 540 | |||
BC117105.1 | - | 2 | 872 | 40126424 | ||
BC117107.1 | - | 2 | 872 | 40126426 | ||
M93292.1 | - | 2 | 836 | |||
M95599.1 | - | 2 | 925 |
