siRNA Details
Gene Symbol: Hoxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010459.7 | NP_034589.3 | 2 | 1602 | ||
GenBank mRNA | AK133945.1 | - | 2 | 1131 | ||
M36654.1 | - | 2 | 1168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
