siRNA Details
Gene Symbol: Kcnj6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010606.2 | NP_034736.2 | 4 | 2105 | ||
GenBank mRNA | AB029502.1 | - | 4 | 1867 | ||
AK139056.1 | - | 4 | 2110 | |||
BC120794.1 | - | 4 | 2015 | 40129717 | ||
BC125565.1 | - | 4 | 2015 | 40130080 | ||
U51123.1 | - | 4 | 2058 | |||
U51124.1 | - | 4 | 2058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
