siRNA Details
Gene Symbol: Klk1b24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010643.1 | NP_034773.1 | 570 | |||
GenBank mRNA | AB039277.1 | - | 570 | |||
AF019086.1 | - | 142 | ||||
BC146015.1 | - | 4 | 597 | 40131318 | ||
BC146019.1 | - | 4 | 597 | 40131322 |
