siRNA Details
Gene Symbol: Krt6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010669.2 | NP_034799.2 | 3 | 833 | ||
GenBank mRNA | AK137688.1 | - | 3 | 832 | ||
BC119554.1 | - | 791 | 40057432 | |||
BC119555.1 | - | 791 | 40057433 | |||
BC122884.1 | - | 3 | 790 | |||
BC124556.1 | - | 3 | 790 | |||
BC139807.1 | - | 791 | 40057436 |
Gene Symbol: Gm5478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC046626.1 | - | 3 | 830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
