siRNA Details
Gene Symbol: Gm10228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270487.1 | NP_001257416.1 | 1 | 377 | ||
GenBank mRNA | AK137757.1 | - | 1 | 378 | ||
D86421.1 | - | 1 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
