siRNA Details
Gene Symbol: Lamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010685.4 | NP_034815.2 | 9 | 3084 | ||
GenBank mRNA | AK032974.1 | - | 9 | 3049 | ||
AK048174.1 | - | 9 | 3026 | |||
AK080234.1 | - | 1 | 1180 | |||
AK088804.1 | - | 10 | 3061 | |||
AK128946.1 | - | 7 | 2746 | |||
AK136550.1 | - | 9 | 3050 | |||
AK150766.1 | - | 9 | 3057 | |||
AK163933.1 | - | 9 | 3085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
