siRNA Details
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010718.4 | NP_034848.1 | 2 | 256 | ||
GenBank mRNA | AB005131.1 | - | 2 | 109 | ||
AB008117.1 | - | 2 | 192 | |||
AK209388.1 | - | 1 | 66 | |||
BC007129.1 | - | 2 | 199 | 3496836 | ||
BY253265.1 | - | 2 | 256 | |||
U88618.1 | - | 2 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
