siRNA Details
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103171.1 | NP_001096641.1 | 2 | 513 | ||
NM_010862.2 | NP_034992.2 | 2 | 513 | |||
GenBank mRNA | AF144095.1 | - | 2 | 513 | ||
AY331132.1 | - | 2 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
