siRNA Details
Gene Symbol: Ncf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286037.1 | NP_001272966.1 | 4 | 339 | ||
NM_010876.4 | NP_035006.3 | 4 | 339 | |||
GenBank mRNA | AB002663.1 | - | 4 | 318 | ||
AK149619.1 | - | 4 | 340 | |||
AK149668.1 | - | 4 | 306 | |||
AK149732.1 | - | 4 | 340 | |||
AK150945.1 | - | 4 | 305 | |||
AK151316.1 | - | 4 | 306 | |||
AK151573.1 | - | 3 | 165 | |||
AK151593.1 | - | 4 | 304 | |||
AK152386.1 | - | 4 | 302 | |||
AK152992.1 | - | 4 | 306 | |||
AK162308.1 | - | 4 | 306 | |||
AK162623.1 | - | 4 | 309 | |||
AK170462.1 | - | 4 | 306 | |||
AK171559.1 | - | 4 | 305 | |||
BC026529.1 | - | 3 | 162 | |||
L11455.1 | - | 4 | 276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
