siRNA Details
Gene Symbol: Neurod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010895.3 | NP_035025.3 | 2 | 1812 | ||
GenBank mRNA | AK158767.1 | - | 2 | 1944 | ||
BC058965.1 | - | 2 | 1761 | 6817440 | ||
CJ237767.1 | - | 1 | 284 | |||
D83507.1 | - | 2 | 1628 |

More Information
Other Species Targeted

Gene Ontology Categories:
