siRNA Details
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286541.1 | NP_001273470.1 | 2, 3 | 359 | ||
NM_001286542.1 | NP_001273471.1 | 2 | 233 | |||
NM_001286551.1 | NP_001273480.1 | 3, 4 | 513 | |||
NM_011014.3 | NP_035144.1 | 3, 4 | 560 | |||
GenBank mRNA | AF004927.1 | - | 3, 4 | 514 | ||
AF030198.1 | - | 3, 4 | 474 | |||
AK002718.1 | - | 3, 4 | 493 | |||
AK154300.1 | - | 3, 4 | 532 | |||
AK159886.1 | - | 3, 4 | 494 | |||
AY390767.1 | - | 2 | 126 | |||
AY390768.1 | - | 2, 3 | 252 | |||
BC002000.1 | - | 3, 4 | 509 | 3484605 | ||
BC019930.1 | - | 3, 4 | 488 | 5123891 | ||
BM507398.1 | - | 3, 4 | 405 |
