siRNA Details
Gene Symbol: Pabpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011033.2 | NP_035163.1 | 1 | 479 | ||
GenBank mRNA | AK077154.1 | - | 1 | 480 | ||
AK131715.1 | - | 1 | 458 | |||
BC051134.1 | - | 1 | 459 | 6704229 | ||
X75959.1 | - | 1 | 455 |
