siRNA Details
Gene Symbol: Pax8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011040.4 | NP_035170.1 | 3 | 334 | ||
GenBank mRNA | AK050717.1 | - | 3 | 310 | ||
AK052610.1 | - | 3 | 310 | |||
BC020526.1 | - | 3 | 301 | 4239835 | ||
X57487.1 | - | 3 | 291 |
