siRNA Details
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286538.1 | NP_001273467.1 | 4 | 567 | ||
NM_001286539.1 | NP_001273468.1 | 3 | 604 | |||
NM_001286540.1 | NP_001273469.1 | 4 | 583 | |||
NM_001286541.1 | NP_001273470.1 | 3 | 496 | |||
NM_001286542.1 | NP_001273471.1 | 2 | 370 | |||
NM_001286551.1 | NP_001273480.1 | 4 | 650 | |||
NM_001286605.1 | NP_001273534.1 | 4 | 520 | |||
NM_011014.3 | NP_035144.1 | 4 | 697 | |||
GenBank mRNA | AF004927.1 | - | 4 | 651 | ||
AF030198.1 | - | 4 | 611 | |||
AF226605.1 | - | 3 | 497 | |||
AK002718.1 | - | 4 | 630 | |||
AK154300.1 | - | 4 | 669 | |||
AK159886.1 | - | 4 | 631 | |||
AY390764.1 | - | 3 | 497 | |||
AY390765.1 | - | 4 | 460 | |||
AY390766.1 | - | 4 | 476 | |||
AY390767.1 | - | 2 | 263 | |||
AY390768.1 | - | 3 | 389 | |||
BC002000.1 | - | 4 | 646 | 3484605 | ||
BC019930.1 | - | 4 | 625 | 5123891 | ||
BM507398.1 | - | 4 | 542 |
