siRNA Details
Gene Symbol: Pira1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011087.1 | NP_035217.1 | 2418 | |||
GenBank mRNA | U96682.1 | - | 2418 |
Gene Symbol: Gm10693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137448.1 | - | 7 | 3021 |
Gene Symbol: Pira2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011089.2 | NP_035219.2 | 10 | 2437 | ||
GenBank mRNA | AK140776.1 | - | 10 | 2443 | ||
AK172046.1 | - | 2438 |
Gene Symbol: Pira6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289428.1 | NP_001276357.1 | 2417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
