siRNA Details
Gene Symbol: Olfr54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010997.1 | NP_035127.1 | 1 | 889 | ||
GenBank mRNA | BC099884.1 | - | 1 | 888 | ||
BC099885.1 | - | 1 | 888 | |||
BC100678.1 | - | 1 | 888 |

More Information
Other Species Targeted

Gene Ontology Categories:
