siRNA Details
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286538.1 | NP_001273467.1 | 4 | 792 | ||
NM_001286539.1 | NP_001273468.1 | 3 | 829 | |||
NM_001286540.1 | NP_001273469.1 | 4 | 808 | |||
NM_001286541.1 | NP_001273470.1 | 3 | 721 | |||
NM_001286542.1 | NP_001273471.1 | 2 | 595 | |||
NM_001286551.1 | NP_001273480.1 | 4 | 875 | |||
NM_001286605.1 | NP_001273534.1 | 4 | 745 | |||
NM_011014.3 | NP_035144.1 | 4 | 922 | |||
GenBank mRNA | AF004927.1 | - | 4 | 876 | ||
AF030198.1 | - | 4 | 836 | |||
AK002718.1 | - | 4 | 855 | |||
AK154300.1 | - | 4 | 893 | |||
AK159886.1 | - | 4 | 855 | |||
AK181288.1 | - | 1 | 102 | |||
AY390765.1 | - | 4 | 685 | |||
AY390767.1 | - | 2 | 488 | |||
AY390768.1 | - | 3 | 614 | |||
BC002000.1 | - | 4 | 870 | 3484605 | ||
BC019930.1 | - | 4 | 850 | 5123891 |
