siRNA Details
Gene Symbol: YES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005433.3 | NP_005424.1 | 2 | 446 | ||
XM_005258139.3 | XP_005258196.1 | 2 | 408 | |||
XM_017025960.1 | XP_016881449.1 | 2 | 288 | |||
GenBank mRNA | AK314063.1 | - | 2 | 390 | ||
BC048960.1 | - | 2 | 442 | 5260751 | ||
M15990.1 | - | 2 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
