siRNA Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011127.2 | NP_035257.1 | 4 | 1713 | ||
NM_175686.3 | NP_783617.2 | 5 | 1785 | |||
GenBank mRNA | AB221686.1 | - | 4 | 698 | ||
AK008112.1 | - | 616 | ||||
AK008428.2 | - | 6 | 615 | |||
BC092372.1 | - | 5 | 1134 | 30533016 | ||
L06502.1 | - | 5 | 950 | |||
U03873.1 | - | 5 | 1791 | |||
X59725.1 | - | 4 | 839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
