siRNA Details
Gene Symbol: Pou2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011137.3 | NP_035267.2 | 17 | 2580 | ||
NM_198932.3 | NP_945150.2 | 16 | 2508 | |||
NM_198933.3 | NP_945151.2 | 16 | 2580 | |||
NM_198934.3 | NP_945152.2 | 17 | 2652 | |||
GenBank mRNA | AF095460.1 | - | 2 | 528 | ||
AF508939.1 | - | 16 | 2489 | |||
AK028237.1 | - | 17 | 2582 | |||
S65461.1 | - | 15 | 2408 | |||
X68362.1 | - | 17 | 2553 | |||
X68363.1 | - | 16 | 2479 | |||
X70324.1 | - | 15 | 2340 | |||
X70325.1 | - | 6 | 1321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
