siRNA Details
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011139.2 | NP_035269.2 | 10 | 1066 | ||
GenBank mRNA | AK153697.1 | - | 1156 | |||
L14677.1 | - | 10 | 1064 | |||
Z18537.1 | - | 10 | 1067 |

More Information
Other Species Targeted

Gene Ontology Categories:
