siRNA Details
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011149.2 | NP_035279.2 | 5 | 690 | ||
GenBank mRNA | AK002357.1 | - | 5 | 637 | ||
AK128977.1 | - | 4 | 3050 | |||
AK154073.1 | - | 5 | 629 | |||
BC013061.1 | - | 5 | 627 | 3483267 | ||
CA321924.1 | - | 3 | 269 | |||
M60456.1 | - | 5 | 609 | |||
X58990.1 | - | 5 | 735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
